missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
44.92 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -116.0°C |
Color | Colorless |
Density | 0.7140g/mL |
Boiling Point | 34.6°C |
Quantity | 1 L |
Flash Point | −45°C |
Packaging | AcroSeal™ Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.3510 to 1.3530 |
Assay Percent Range | 99.5% |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
Product Identifier
- Diethyl ether
- Danger
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
- Flammable liquid Category 1
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P403+P235-Store in a well-ventilated place. Keep cool.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'