missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals

Product Code. 13167754
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
Glass Bottle
Metal Drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116.0°C
Density 0.7140g/mL
Boiling Point 34.6°C
Quantity 25 L
Flash Point −45°C
Infrared Spectrum Authentic
Packaging Metal Drum
Physical Form Liquid
Refractive Index 1.3510 to 1.3530
Assay Percent Range 99+%
Residue after Evaporation 0.002 % max.
Specific Gravity 0.714
Linear Formula (CH3CH2)2O
UN Number 1155
Viscosity 0.2448 mPa.s (20°C)
Water 0.2% Max. (Coulometric)
Solubility Information Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid, miscible with lower aliphatic alcohols, benzene,, chloroform, petroleum ether and many oils
Formula Weight 74.12
Percent Purity ≥99%
Grade Pure
Chemical Name or Material Diethyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 1
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'