missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, ACS reagent, anhydrous, contains BHT as inhibitor, ≥99.0%, Honeywell Riedel-de Haën™

ACS reagent, anhydrous, contains BHT as inhibitor,≥99.0%

£75.00 - £1452.67

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15626680
View Documents
Honeywell-Riedel-de Haen
673811-250ML
250 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15616680
View Documents
Honeywell-Riedel-de Haen
673811-1L
1 L Aluminum Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15668440
View Documents Promotion Details
Honeywell-Riedel-de Haen
673811-4L
4 L Aluminum Bottle
£389.49
4 L
Please to purchase this item. Need a web account? Register with us today!
15636680
View Documents
Honeywell-Riedel-de Haen
673811-4X4L
4 x 4 L Glass Bottle
£1452.67
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

60-29-7
74.12
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC

Specifications

-116°C (literature)
≤100.0000%
34°C to 35°C
anhydrous
(CH3CH2)2O
UN1155
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CCOCC
74.12
CHEBI:35702
>99%
Diethyl ether, ACS reagent, anhydrous, contains BHT as inhibitor, ≥99.0%
60-29-7
0.706 g/mL at 25 °C(lit.)
−45°C
C4H10O
MFCD00011646
1696894
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
ACS Reagent
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 1260 days from date of manufacture

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether

missing translation for 'einecsNumber' : 200-467-2

Recommended Storage : Room Temp

SDS
Documents

Documents

Product Certifications