missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl ether, ACS Reagent, contains ∼2% ethanol and ∼10ppm BHT as inhibitor, ≥98.0%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Melting Point | -116°C (literature) |
Density | 0.706 g/mL at 25 °C(lit.) |
Boiling Point | 34°C to 35°C |
Quantity | 1 L |
Flash Point | −45°C |
Packaging | Glass Bottle |
Linear Formula | (CH3CH2)2O |
UN Number | UN1155 |
Beilstein | 1696894 |
Formula Weight | 74.12 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'