missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, Anhydrous, BAKER, J.T. Baker™
316.35 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
5 L
Packaging:
EcoTainer Metal Safety Can
Description
A grade of chemicals for laboratory use, for organic synthesis, and for routine research usually controlled by physical criteria and often by assay (by GC).
Specifications
Melting Point | -116.3°C |
CAS | 60-29-7 |
Color | Colorless |
Acidity | Passes test |
Molecular Formula | C4H10O |
Linear Formula | (C2H5)2O |
MDL Number | MFCD00011646 |
Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
Solubility Information | 12g/L in water |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'