missing translation for 'onlineSavingsMsg'
Learn More
Diethyl Ether, Certified AR for Analysis, Stabilised with BHT, meets analytical specification of Ph.Eur, Fisher Chemical™ Product Code.: 11323440

Diethyl Ether, Certified AR for Analysis, Stabilised with BHT, meets analytical specification of Ph.Eur, Fisher Chemical™

Product Code. 11323440
25 L, Metal Can
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Aluminum Can
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Product Code. 11323440

Brand: Fisher Chemical D/2450/25

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

C4H10O, CAS Number-60-29-7, ether, pronarcol, 3-oxapentane, anesthetic ether, aether, anaesthetic ether, diethyl oxide, diethyl ether, ethyl ether, ethyl oxide, 2.5L, 34.6 deg.C, CHEBI:35702, Colorless, 74.12g/mol, RTZKZFJDLAIYFH-UHFFFAOYSA-N, ethoxyethane, -116 deg.C, 11646, 74.123

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Spécification

Melting Point -116°C
Color Colorless
Boiling Point 34.6°C
Quantity 25 L
Packaging Metal Can
Physical Form Liquid
Vapor Pressure 587mbar at 20°C
Viscosity 0.2448cp at 20°C
Formula Weight 74.12g/mol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2527135 Certificate TypeCertificate of Analysis Date19/02/2025 Catalog NumberD/2450/17Z,D/2450/15,D/2450/15Z,D/2450/25,D/2450/PB08,D/2450/27,D/2450/PB15,D/2450/PC15,D/2450/PB17,D/2450/MS21
compliance-icons compliance-icons
Identifiant produit
  • Diethyl ether
Mention d'avertissement
  • Danger
Catégorie de danger
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Mention de danger
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
  • EUH019-May form explosive peroxides.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
Diethyl Ether, Certified AR for Analysis, Stabilised with BHT, meets analytical specification of Ph.Eur, Fisher Chemical™ >

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis