missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, CHROMASOLV™, for HPLC, inhibitor-free, ≥99.9%, Honeywell™ Riedel-de Haen™

CHROMASOLV™, for HPLC, inhibitor-free,≥99.9%

£36.28 - £500.79

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15653150
View Documents
Honeywell-Chromasolv
309966-100ML
100 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15663150
View Documents
Honeywell-Chromasolv
309966-1L
1 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15673150
View Documents
Honeywell-Chromasolv
309966-6X1L
6 x 1 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

 

Specifications

Chemical Identifiers

60-29-7
74.12
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC

Specifications

-116°C (literature)
60-29-7
0.706 g/mL at 25 °C(lit.)
−45°C
(CH3CH2)2O
UN1155
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CCOCC
74.12
CHEBI:35702
>99.9%
Diethyl ether, CHROMASOLV™, for HPLC, inhibitor-free, ≥99.9%
max. 1.00 at 218 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.04 at 275 nm, max. 0.20 at 250 nm
≤100.0000%
34°C to 35°C
C4H10O
MFCD00011646
1696894
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
HPLC
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 1260 days from date of manufacture

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether

missing translation for 'einecsNumber' : 200-467-2

Recommended Storage : Room Temp

Product Identifier
  • Diethyl ether

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3

Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.