missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl ether, For pesticide residue analysis, Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Specifications
Color | Colorless |
Linear Formula | (CH3CH2)2O |
UN Number | 1155 |
Beilstein | 1696894 |
Formula Weight | 74.12g/mol |
Grade | CHROMASOLV™ |
Density | 0.706g/mL at 25°C |
Quantity | 1 L |
Explosion Limit | 0.365 |
Flash Point | −40°C (−40°F) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'