Learn More
Thermo Scientific Chemicals Diethyl ether, HPLC Grade, 99%, stab. with ethanol
Widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents.
£68.80 - £220.00
Specifications
Chemical Name or Material | Diethyl ether |
---|---|
CAS | 60-29-7 |
Physical Form | Liquid |
Molecular Formula | C4H10O |
MDL Number | MFCD00011646 |
Description
It is a widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethyl ether | |
Liquid | |
MFCD00011646 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12 |
60-29-7 | |
C4H10O | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CCOCC | |
74.12 | |
CHEBI:35702 |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P314-P330-P370+P378q-P501c
H224-H302-H305-H335-H336-H373
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: I
missing translation for 'einecsNumber' : 200-467-2
missing translation for 'rtecsNumber' : KI5775000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only