missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, puriss., ≥99.5% (GC), Honeywell™ Riedel-de Haën™
Puriss., meets analytical specification of BP, Ph. Eur.,≥99.5% (GC)
£108.25 - £1023.66
Chemical Identifiers
CAS | 60-29-7 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | MFCD00011646 |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
PubChem CID | 3283 |
ChEBI | CHEBI:35702 |
IUPAC Name | ethoxyethane |
SMILES | CCOCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15632460
|
Honeywell-Riedel-de Haen
24004-2.5L-ALU |
2.5 L | Aluminum Bottle | N/A | |||||
15602470
|
Honeywell-Riedel-de Haen
24004-6X1L |
6 x 1 L | Glass Bottle |
£359.24
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15662460
|
Honeywell-Riedel-de Haen
24004-4X2.5L |
4 x 2.5 L | Glass Bottle |
£543.13
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15672460
|
Honeywell-Riedel-de Haen
24004-4X5L |
4 x 5 L | Aluminum Bottle |
£1023.66
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Chemical Identifiers
60-29-7 | |
74.12 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
3283 | |
ethoxyethane |
C4H10O | |
MFCD00011646 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CHEBI:35702 | |
CCOCC |
Specifications
-116°C | |
0.706g/mL at 25°C | |
0.365 | |
320°F | |
C4H10O | |
MFCD00011646 | |
1696894 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12g/mol | |
puriss. |
60-29-7 | |
34.6°C | |
−45°C (−49°F) | |
Liquid | |
(CH3CH2)2O | |
UN1155 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
≥99.5% (GC) | |
Diethyl Ether, Meets analytical specification of Ph.Eur.BP |
Safety and Handling
P210-P261
H224-H302-H336
missing translation for 'dotInformation' : class 3 - PG 1 - Diethyl ether
missing translation for 'einecsNumber' : 200-467-2
missing translation for 'rtecsNumber' : KI5775000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Diethyl ether
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.