missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, puriss., Honeywell™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
88.83 GBP valid until 2024-08-31
Use promo code "21641" to get your promotional price.
Use promo code "21641" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
135 kg
240 L
Specifications
Density | 0.706g/mL at 25°C |
Quantity | 2.5 L |
Explosion Limit | 0.365 |
Flash Point | −40°C (−40°F) |
Ignition Point | 320°F |
Linear Formula | (CH3CH2)2O |
UN Number | 1155 |
Beilstein | 1696894 |
Formula Weight | 74.12g/mol |
Percent Purity | ≥99.5% (GC) |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Diethyl ether
- Danger
- Flammable liquid Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'