missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, puriss. p.a. ACS Reagent, Honeywell™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
27 L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Density | 0.706g/mL at 25°C |
Quantity | 27 L |
Explosion Limit | 0.365 |
Flash Point | −40°C (−40°F) |
Ignition Point | 320°F |
Linear Formula | (CH3CH2)2O |
UN Number | 1155 |
Beilstein | 1696894 |
Formula Weight | 74.12g/mol |
Percent Purity | ≥99.8% |
Show More |
Safety and Handling
Product Identifier
- Diethyl ether
- Danger
- Flammable liquid Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'