missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, puriss. p.a. ACS Reagent, Honeywell™

Puriss. p.a., contains BHT as inhibitor, ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%

£27.15 - £777.27

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15673680
View Documents
Honeywell Riedel-de Haën
32203-500ML
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15623680
View Documents Promotion Details
Honeywell Riedel-de Haën
32203-1L
1 L
£62.65
1 L
Please to purchase this item. Need a web account? Register with us today!
15633680
View Documents
Honeywell Riedel-de Haën
32203-2.5L
2.5 L
£110.83
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15683680
View Documents
Honeywell Riedel-de Haën
32203-5L
5 L
£249.48
5 L
Please to purchase this item. Need a web account? Register with us today!
15643680
View Documents
Honeywell Riedel-de Haën
32203-27L
27 L
£777.27
27 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

60-29-7
74.12
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC

Specifications

60-29-7
0.365
320°F
(CH3CH2)2O
1155
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CCOCC
74.12
CHEBI:35702
≥99.8%
Diethyl Ether, Reag. Ph. Eur. reag. ISO
0.706g/mL at 25°C
−40°C (−40°F)
C4H10O
MFCD00011646
1696894
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12g/mol
ACS/puriss. p.a.
Videos
Safety and Handling

Safety and Handling

P210-P261

H224-H302-H336

Danger

missing translation for 'einecsNumber' : 200-467-2

missing translation for 'rtecsNumber' : KI5775000

Product Identifier
  • Diethyl ether

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3

Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications