missing translation for 'onlineSavingsMsg'
Learn More

Diethyl fumarate, 98%

Product Code. p-108993 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
250 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Unit Size:
10g
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 10773641

Brand: Thermo Scientific Acros 114380100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-91-6 | C8H12O4 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-91-6
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00064455
InChI Key IEPRKVQEAMIZSS-AATRIKPKSA-N
Synonym diethyl fumarate, fumaric acid, diethyl ester, anti-psoriaticum, diethyl 2e-but-2-enedioate, fumaric acid diethyl ester, diethyl e-but-2-enedioate, 2-butenedioic acid e-, diethyl ester, trans-2-butenedioic acid diethyl ester, 2-butenedioic acid 2e-, diethyl ester, unii-5wbu5a3e8a
PubChem CID 638144
ChEBI CHEBI:87388
IUPAC Name diethyl (E)-but-2-enedioate
SMILES CCOC(=O)\C=C\C(=O)OCC

Specifications

Melting Point 1.0°C to 2.0°C
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 218.0°C to 219.0°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula C2H5O2CCH=CHCO2C2H5
Refractive Index 1.4396 to 1.4416
Quantity 10 g
Specific Gravity 1.05
Solubility Information Solubility in water: immiscible.
Formula Weight 172.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl fumarate, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.