Learn More
Diethyl hexafluoroglutarate, 97%, Thermo Scientific Chemicals
£15.67 - £67.90
Chemical Identifiers
CAS | 424-40-8 |
---|---|
Molecular Formula | C9H10F6O4 |
Molecular Weight (g/mol) | 296.17 |
MDL Number | MFCD00015156 |
InChI Key | MSDPXVBLFJODJO-UHFFFAOYSA-N |
Synonym | diethyl hexafluoroglutarate, diethyl perfluoroglutarate, diethyl hexafluoropentanedioate, hexafluoroglutaric acid diethyl ester, pentanedioic acid, hexafluoro-, diethyl ester, glutaric acid, hexafluoro-, diethyl ester, hexafluoropentanedioic acid diethyl ester, diethylester kyseliny hexafluorglutarove, diethylester kyseliny hexafluorglutarove czech, pentanedioic acid, 2,2,3,3,4,4-hexafluoro-, 1,5-diethyl ester |
PubChem CID | 9874 |
SMILES | CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(=O)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11420567
|
Thermo Scientific Alfa Aesar
B22428.03 |
1 g | |||||||
11430567
|
Thermo Scientific Alfa Aesar
B22428.06 |
5 g |
£67.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
424-40-8 | |
296.17 | |
MSDPXVBLFJODJO-UHFFFAOYSA-N | |
9874 |
C9H10F6O4 | |
MFCD00015156 | |
diethyl hexafluoroglutarate, diethyl perfluoroglutarate, diethyl hexafluoropentanedioate, hexafluoroglutaric acid diethyl ester, pentanedioic acid, hexafluoro-, diethyl ester, glutaric acid, hexafluoro-, diethyl ester, hexafluoropentanedioic acid diethyl ester, diethylester kyseliny hexafluorglutarove, diethylester kyseliny hexafluorglutarove czech, pentanedioic acid, 2,2,3,3,4,4-hexafluoro-, 1,5-diethyl ester | |
CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(=O)OCC |
Specifications
424-40-8 | |
224°C | |
1.36 | |
1804802 | |
MSDPXVBLFJODJO-UHFFFAOYSA-N | |
diethyl 2,2,3,3,4,4-hexafluoropentanedioate | |
9874 | |
97% |
1.365 | |
C9H10F6O4 | |
MFCD00015156 | |
diethyl hexafluoroglutarate, diethyl perfluoroglutarate, diethyl hexafluoropentanedioate, hexafluoroglutaric acid diethyl ester, pentanedioic acid, hexafluoro-, diethyl ester, glutaric acid, hexafluoro-, diethyl ester, hexafluoropentanedioic acid diethyl ester, diethylester kyseliny hexafluorglutarove, diethylester kyseliny hexafluorglutarove czech, pentanedioic acid, 2,2,3,3,4,4-hexafluoro-, 1,5-diethyl ester | |
CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(=O)OCC | |
296.17 | |
296.16 | |
Diethyl hexafluoroglutarate |
Safety and Handling
GHS H Statement
H332-H315-H319-H335-H227
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H332-H335
missing translation for 'einecsNumber' : 207-035-2
missing translation for 'rtecsNumber' : MA3750500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only