missing translation for 'onlineSavingsMsg'
Learn More

Diethyl isobutylmalonate, 98%, Thermo Scientific Chemicals

Product Code. 11327286
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10203-58-4
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00026869
InChI Key OFRFGNSZCYDFOH-UHFFFAOYSA-N
Synonym diethyl isobutylmalonate, isobutylmalonic acid diethyl ester, diethyl 2-isobutylmalonate, propanedioic acid, 2-methylpropyl-, diethyl ester, malonic acid, isobutyl-, diethyl ester, diethyl 2-2-methylpropyl propanedioate, ethyl 2-carbethoxy-4-methyl pentanoate, propanedioic acid, 2-2-methylpropyl-, 1,3-diethyl ester, 1,3-diethyl 2-2-methylpropyl propanedioate, diethylisobutylmalonate
PubChem CID 25047
IUPAC Name diethyl 2-(2-methylpropyl)propanedioate
SMILES CCOC(=O)C(CC(C)C)C(=O)OCC

Specifications

Density 0.97
Boiling Point 225°C
Flash Point >100°C (212°F)
Refractive Index 1.422
Quantity 100 g
Beilstein 1103030
Formula Weight 216.28
Percent Purity 98%
Chemical Name or Material Diethyl isobutylmalonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'