Learn More
Diethyl maleate, 97%, Thermo Scientific Chemicals
£19.30 - £58.40
Chemical Identifiers
CAS | 141-05-9 |
---|---|
Molecular Weight (g/mol) | 172.18 |
MDL Number | MFCD00009191 |
InChI Key | IEPRKVQEAMIZSS-WAYWQWQTSA-N |
Synonym | diethyl maleate, ethyl maleate, maleic acid, diethyl ester, diethylmaleate, maleic acid diethyl ester, 2-butenedioic acid z-, diethyl ester, diethyl z-but-2-enedioate, diethyl 2z-but-2-enedioate, unii-g81wqb56ol, ccris 941 |
PubChem CID | 5271566 |
ChEBI | CHEBI:68508 |
IUPAC Name | diethyl (Z)-but-2-enedioate |
SMILES | CCOC(=O)C=CC(=O)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10234000
|
Thermo Scientific Acros
114440050 |
5 g | Glass bottle |
£19.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10419220
|
Thermo Scientific Acros
114442500 |
250 g | Glass bottle |
£21.40
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10336370
|
Thermo Scientific
114440010 |
1 kg | Glass bottle |
£58.40
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-05-9 | |
MFCD00009191 | |
diethyl maleate, ethyl maleate, maleic acid, diethyl ester, diethylmaleate, maleic acid diethyl ester, 2-butenedioic acid z-, diethyl ester, diethyl z-but-2-enedioate, diethyl 2z-but-2-enedioate, unii-g81wqb56ol, ccris 941 | |
CHEBI:68508 | |
CCOC(=O)C=CC(=O)OCC |
172.18 | |
IEPRKVQEAMIZSS-WAYWQWQTSA-N | |
5271566 | |
diethyl (Z)-but-2-enedioate |
Specifications
141-05-9 | |
1.0600g/mL | |
93°C | |
96% min. (GC) | |
C2H5O2CCH=CHCO2C2H5 | |
MFCD00009191 | |
15,3139 | |
diethyl maleate, ethyl maleate, maleic acid, diethyl ester, diethylmaleate, maleic acid diethyl ester, 2-butenedioic acid z-, diethyl ester, diethyl z-but-2-enedioate, diethyl 2z-but-2-enedioate, unii-g81wqb56ol, ccris 941 | |
IEPRKVQEAMIZSS-WAYWQWQTSA-N | |
diethyl (Z)-but-2-enedioate | |
5271566 | |
172.18 | |
Liquid |
-10.0°C | |
225.0°C | |
Authentic | |
C8H12O4 | |
1.4380 to 1.4420 | |
02,751 | |
1.06 | |
Solubility in water: insoluble. Other solubilities: readily soluble in alcohol, diethyl ether | |
CCOC(=O)C=CC(=O)OCC | |
172.18 | |
CHEBI:68508 | |
97% | |
Diethyl maleate, ca.90% |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
GHS P Statement:
Wear eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-451-9
missing translation for 'rtecsNumber' : ON1225000
missing translation for 'tsca' : TSCA
- Diethyl maleate
Signal Word
- Warning
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only