missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl malonate, 99+%, Thermo Scientific Chemicals
£16.80 - £281.00
Chemical Identifiers
CAS | 105-53-3 |
---|---|
Molecular Formula | C7H12O4 |
Molecular Weight (g/mol) | 160.17 |
InChI Key | IYXGSMUGOJNHAZ-UHFFFAOYSA-N |
Synonym | diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van |
PubChem CID | 7761 |
IUPAC Name | diethyl propanedioate |
SMILES | CCOC(=O)CC(=O)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10070760
|
Thermo Scientific Acros
114450250 |
25 g | Glass bottle |
£16.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10599240
|
Thermo Scientific Acros
114455000 |
500 g | Glass bottle |
£30.40
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10192280
|
Thermo Scientific Acros
114450010 |
1 kg | Glass bottle |
£50.10
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10073943
|
Thermo Scientific Acros
114450025 |
2.5 kg | Glass bottle |
£80.80
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11935021
|
Thermo Scientific Acros
114450100 |
10 kg | Plastic drum |
£281.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
105-53-3 | |
160.17 | |
diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van | |
diethyl propanedioate |
C7H12O4 | |
IYXGSMUGOJNHAZ-UHFFFAOYSA-N | |
7761 | |
CCOC(=O)CC(=O)OCC |
Specifications
105-53-3 | |
1.0550g/mL | |
93°C | |
99% min. (GC) | |
C2H5O2CCH2CO2C2H5 | |
02,573 | |
15,3878 | |
diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van | |
IYXGSMUGOJNHAZ-UHFFFAOYSA-N | |
diethyl propanedioate | |
7761 | |
160.17 | |
Liquid |
-50.0°C | |
199.0°C | |
Authentic | |
C7H12O4 | |
1.4130 to 1.4150 | |
10,182; 13,69 | |
1.055 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in ethanol, ether and acetone | |
CCOC(=O)CC(=O)OCC | |
160.17 | |
2.14 mPa.s (10°C) | |
99+% | |
Diethyl malonate |
Safety and Handling
missing translation for 'einecsNumber' : 203-305-9
RUO – Research Use Only