missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phenyl orthoformate, 97%, Thermo Scientific Chemicals

Product Code. 11427044
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14444-77-0
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.246
MDL Number MFCD00009228
InChI Key QTURWMMVIIBRRP-UHFFFAOYSA-N
Synonym diethyl phenyl orthoformate, diethoxymethoxy benzene, benzene, diethoxymethoxy, diethoxymethoxy-benzene, acmc-209csr, diethoxy phenoxy methane, diethoxymethoxy benzene #, diethylphenyl orthoformate, orthoformic acid diethyl phenyl ester
PubChem CID 84440
IUPAC Name diethoxymethoxybenzene
SMILES CCOC(OCC)OC1=CC=CC=C1

Specifications

Melting Point -26°C
Density 1.015
Boiling Point 103°C to 104°C (10 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.4795
Quantity 10 g
Beilstein 1872634
Sensitivity Moisture sensitive
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material Diethyl phenyl orthoformate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'