missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phenylmalonate, 98%, Thermo Scientific Chemicals

£14.94 - £158.00

Chemical Identifiers

CAS 83-13-6
MDL Number MFCD00009144
InChI Key FGYDHYCFHBSNPE-UHFFFAOYSA-N
Synonym diethyl phenylmalonate, diethyl 2-phenylmalonate, phenylmalonic acid diethyl ester, propanedioic acid, phenyl-, diethyl ester, diethylphenylmalonate, 1,3-diethyl 2-phenylpropanedioate, malonic acid, phenyl-, diethyl ester, propanedioic acid, 2-phenyl-, 1,3-diethyl ester, diethyl 2-phenylpropane-1,3-dioate, diethyl-phenylmalonate
PubChem CID 66514
IUPAC Name diethyl 2-phenylpropanedioate
SMILES CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10193430
View Documents
Thermo Scientific Acros
150260050
5 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10001620
View Documents
Thermo Scientific Acros
150261000
100 mL Glass bottle
£18.90
100mL
Please to purchase this item. Need a web account? Register with us today!
10376980
View Documents
Thermo Scientific Acros
150262500
250 mL Glass bottle
£34.70
250mL
Please to purchase this item. Need a web account? Register with us today!
10418300
View Documents
Thermo Scientific Acros
150260010
1 L Glass bottle
£89.00
1 L
Please to purchase this item. Need a web account? Register with us today!
10751925
View Documents
Thermo Scientific Acros
150260025
2.5 L Glass bottle
£158.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

83-13-6
FGYDHYCFHBSNPE-UHFFFAOYSA-N
66514
CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC
MFCD00009144
diethyl phenylmalonate, diethyl 2-phenylmalonate, phenylmalonic acid diethyl ester, propanedioic acid, phenyl-, diethyl ester, diethylphenylmalonate, 1,3-diethyl 2-phenylpropanedioate, malonic acid, phenyl-, diethyl ester, propanedioic acid, 2-phenyl-, 1,3-diethyl ester, diethyl 2-phenylpropane-1,3-dioate, diethyl-phenylmalonate
diethyl 2-phenylpropanedioate

Specifications

83-13-6
Colorless to Yellow
170.0°C to 172.0°C (14.0 mmHg)
Authentic
C13H16O4
1.4903 to 1.4923
09, 854
diethyl phenylmalonate, diethyl 2-phenylmalonate, phenylmalonic acid diethyl ester, propanedioic acid, phenyl-, diethyl ester, diethylphenylmalonate, 1,3-diethyl 2-phenylpropanedioate, malonic acid, phenyl-, diethyl ester, propanedioic acid, 2-phenyl-, 1,3-diethyl ester, diethyl 2-phenylpropane-1,3-dioate, diethyl-phenylmalonate
FGYDHYCFHBSNPE-UHFFFAOYSA-N
diethyl 2-phenylpropanedioate
66514
98%
Diethyl phenylmalonate
16.0°C to 17.0°C
1.0900g/mL
>112°C
97.5% min. (GC)
C2H5O2CCH(C6H5)CO2C2H5
MFCD00009144
1.09
Solubility in water: immiscible
CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC
236.27
236.27
Liquid After Melting
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 201-456-5

Product Identifier
  • Diethyl phenylmalonate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only