missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 84-66-2 |
---|---|
Molecular Formula | C12H14O4 |
Molecular Weight (g/mol) | 222.24 |
MDL Number | MFCD00009111 |
InChI Key | FLKPEMZONWLCSK-UHFFFAOYSA-N |
Synonym | diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e |
PubChem CID | 6781 |
ChEBI | CHEBI:34698 |
IUPAC Name | diethyl benzene-1,2-dicarboxylate |
SMILES | CCOC(=O)C1=CC=CC=C1C(=O)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10376080
|
Thermo Scientific
114520250 |
25 mL | Glass bottle |
£16.18
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10569410
|
Thermo Scientific Acros
114520010 |
1 L | Glass bottle |
£49.14
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10714801
|
Thermo Scientific Acros
114520025 |
2.5 L | Glass bottle |
£87.84
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10443825
|
Thermo Scientific Acros
114520100 |
10 L | Plastic drum |
£219.09
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
84-66-2 | |
222.24 | |
FLKPEMZONWLCSK-UHFFFAOYSA-N | |
6781 | |
diethyl benzene-1,2-dicarboxylate |
C12H14O4 | |
MFCD00009111 | |
diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e | |
CHEBI:34698 | |
CCOC(=O)C1=CC=CC=C1C(=O)OCC |
Specifications
84-66-2 | |
298.0°C to 299.0°C | |
Authentic | |
C12H14O4 | |
MFCD00009111 | |
15,7483 | |
diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e | |
FLKPEMZONWLCSK-UHFFFAOYSA-N | |
diethyl benzene-1,2-dicarboxylate | |
6781 | |
12 mPa.s (20°C) | |
99% | |
Diethyl phthalate, 99% |
1.1180g/mL | |
156°C | |
98.5% min. (GC) | |
1.5010 to 1.5030 | |
09,798 | |
1.118 | |
Solubility in water: 1g/L (20°C). Other solubilities: soluble in ethanol,diethyl ether,acetone,,benzene,chloroform,carbon tetrachloride | |
CCOC(=O)C1=CC=CC=C1C(=O)OCC | |
222.24 | |
CHEBI:34698 | |
222.24 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 201-550-6
missing translation for 'rtecsNumber' : TI1050000
missing translation for 'tsca' : TSCA
RUO – Research Use Only