missing translation for 'onlineSavingsMsg'
Learn More

Diethyl sulfite, 98%, Thermo Scientific™

Product Code. 10402791
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 623-81-4
Color Colorless
Density 1.0800g/mL
Boiling Point 158°C to 160°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H10O3S
Linear Formula (C2H5O)2SO
Refractive Index 1.4130 to 1.415
MDL Number MFCD00009098
Quantity 10 g
Beilstein 01, 325
Specific Gravity 1.08
Synonym sulfurous acid, diethyl ester, ethyl sulfite, diethyl sulphite, ethyl sulfite, et2so3, diethylsulfite, ethoxysulfinyl ethoxy, diethylsulfit, sulfurous acid diethyl, diethyl sulfite, sulfurous acid diethyl ester
InChI Key NVJBFARDFTXOTO-UHFFFAOYSA-N
SMILES CCOS(=O)OCC
IUPAC Name diethyl sulfite
Molecular Weight (g/mol) 138.18
PubChem CID 12197
Formula Weight 138.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl sulfite, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl sulfite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.