missing translation for 'onlineSavingsMsg'
Learn More

Diethylaluminium iodide, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10586943
Click to view available options
Quantity:
100 mL
800 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2040-00-8, 108-88-3
Molecular Formula C4H10AlI
Molecular Weight (g/mol) 212.01
MDL Number MFCD00001025
InChI Key PPQUYYAZSOKTQD-UHFFFAOYSA-M
Synonym aluminum, diethyliodo, diethylaluminum iodide, diethylaluminium iodide, diethyl iodo alumane, et2ali
PubChem CID 74874
IUPAC Name diethyl(iodo)alumane
SMILES CC[Al](CC)I

Specifications

Color Colorless
Linear Formula (C2H5)2AlI
Quantity 800 mL
Solubility Information Solubility in water: reacts
Formula Weight 212.01
Physical Form Liquid
Chemical Name or Material Diethylaluminium iodide, 0.8M Solution in Toluene
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethylaluminium iodide, 0.9M solution in toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.