missing translation for 'onlineSavingsMsg'
Learn More
Diethylamine, 99.5%, for analysis, Thermo Scientific Chemicals Product Code.: 15572333

Diethylamine, 99.5%, for analysis, Thermo Scientific Chemicals

Product Code. 15572333
250 mL
Click to view available options
Quantity:
50 mL
250 mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15572333

Brand: Thermo Scientific Acros 458492500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-89-7
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00009032
InChI Key HPNMFZURTQLUMO-UHFFFAOYSA-N
Synonym diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamineShow MoreShow Less
PubChem CID 8021
ChEBI CHEBI:85259
IUPAC Name N-ethylethanamine
SMILES CCNCC

Specifications

Melting Point -50.0°C
Density 0.7100g/mL
Boiling Point 55.0°C
Flash Point −23°C
Assay Percent Range 99.4% min. (GC)
Linear Formula (C2H5)2NH
Refractive Index 1.3840 to 1.3860
Quantity 250 mL
Specific Gravity 0.71
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and most common organic, solvents
Viscosity 0.34 mPa.s (25°C)
Formula Weight 73.13
Percent Purity 99.5%
Grade Analysis
Chemical Name or Material Diethylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463444 Certificate TypeCertificate of Analysis Date31/05/2024 Catalog Number458490500,458492500
Lot NumberA0452756 Certificate TypeCertificate of Analysis Date28/04/2023 Catalog Number458490500,458492500
Lot NumberA0393912 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number458490500,458492500
Lot NumberA0393913 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number458490500,458492500
Lot Number1725305 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number458490500,458492500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Diethylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethylamine, 99.5%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.