missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine, 99+%, extra pure, Thermo Scientific Chemicals

£14.90 - £255.27

Chemical Identifiers

CAS 109-89-7
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00009032
InChI Key HPNMFZURTQLUMO-UHFFFAOYSA-N
Synonym diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine
PubChem CID 8021
ChEBI CHEBI:85259
SMILES CCNCC
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10001510
SDS View Documents
Thermo Scientific Acros
149450050
5 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10093073
SDS View Documents
Thermo Scientific Acros
149452500
250 mL Glass bottle
£26.23
250mL
Please to purchase this item. Need a web account? Register with us today!
10636922
SDS View Documents
Thermo Scientific Acros
149450010
1 L Glass bottle
£44.52
1 L
Please to purchase this item. Need a web account? Register with us today!
10001500
SDS View Documents
Thermo Scientific Acros
149450025
2.5 L Glass Bottle
£84.32
2.50 L
Please to purchase this item. Need a web account? Register with us today!
11975161
SDS View Documents
Thermo Scientific Acros
149450100
10 L Metal drum
£255.27
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethylamine, 99+%, extra pure, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.