Learn More
Diethylamine, 99+%, Thermo Scientific Chemicals
£21.00 - £53.80
Chemical Identifiers
CAS | 109-89-7 |
---|---|
Molecular Formula | C4H11N |
Molecular Weight (g/mol) | 73.14 |
MDL Number | MFCD00009032 |
InChI Key | HPNMFZURTQLUMO-UHFFFAOYSA-N |
Synonym | diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine |
PubChem CID | 8021 |
ChEBI | CHEBI:85259 |
SMILES | CCNCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11324499
|
Thermo Scientific Alfa Aesar
A11716.AE |
100 mL |
£21.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449233
|
Thermo Scientific Alfa Aesar
A11716.AP |
500 mL |
£24.90
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459233
|
Thermo Scientific Alfa Aesar
A11716.0F |
2500 mL |
£53.80
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Diethylamine is used as a corrosion inhibitor. It is also used in the production of rubber, resins, dyes and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethylamine is used as a corrosion inhibitor. It is also used in the production of rubber, resins, dyes and pharmaceuticals. It is also used to cook lysergic acid diethylamide (LSD) and is strictly watched by Drug Enforcement Administration (DEA).
Solubility
Miscible with water, alcohol, ether, chloroform and carbon terachloride.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
109-89-7 | |
73.14 | |
HPNMFZURTQLUMO-UHFFFAOYSA-N | |
8021 | |
CCNCC |
C4H11N | |
MFCD00009032 | |
diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine | |
CHEBI:85259 |
Specifications
109-89-7 | |
0.706 | |
-28°C (-18°F) | |
C4H11N | |
MFCD00009032 | |
Air Sensitive | |
Miscible with water,alcohol,ether,chloroform and carbon terachloride. | |
CCNCC | |
73.14 | |
CHEBI:85259 | |
Liquid |
°C to 50°C | |
54°C to 55°C | |
≥99% | |
1.385 | |
605268 | |
diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine | |
HPNMFZURTQLUMO-UHFFFAOYSA-N | |
N-ethylethanamine | |
8021 | |
73.14 | |
Diethylamine |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H332-H311-H314-H335
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II
missing translation for 'einecsNumber' : 203-716-3
missing translation for 'rtecsNumber' : HZ8750000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Air Sensitive
Recommended Storage : Ambient temperatures; Store under Argon
- Diethylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Diethylamine
RUO – Research Use Only