missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylamine, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
Description
Diethylamine, Extra Pure, SLR
Specifications
Specifications
Melting Point | -50°C |
Color | Colorless |
pH | 12 |
Boiling Point | 55°C |
Packaging | Amber glass bottle |
Quantity | 500 mL |
Formula Weight | 73.13g/mol |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Diethylamine
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'