missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals

£33.40 - £252.00

Chemical Identifiers

CAS 660-68-4
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.597
MDL Number MFCD00012499
InChI Key HDITUCONWLWUJR-UHFFFAOYSA-N
Synonym diethylamine hydrochloride, diethylammonium chloride, diethyl amine hydrochloride, n-ethylethanamine hydrochloride, ethanamine, n-ethyl-, hydrochloride, unii-ze9v3g1135, ethanamine, n-ethyl-, hydrochloride 1:1, diethylaminehydrochloride, diethylamine hcl, diethyl amine hcl
PubChem CID 10197650
IUPAC Name N-ethylethanamine;hydrochloride
SMILES CCNCC.Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11480214
View Documents
Thermo Scientific Alfa Aesar
A13637.30
250 g
£33.40
250g
Please to purchase this item. Need a web account? Register with us today!
11470214
View Documents
Thermo Scientific Alfa Aesar
A13637.0B
1000 g
£88.00
1000g
Please to purchase this item. Need a web account? Register with us today!
11490214
View Documents
Thermo Scientific Alfa Aesar
A13637.0I
5000 g
£252.00
5000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Diethylamine hydrochloride acts as a precursor of atrazine and lysergic acid diethylamide. It is also used as a reactant in the production of dyes and pharmaceutical compounds such as ranitidine. Further, it is used in Mannich reaction with paraformaldehyde. In addition to this, it is employed in the synthesis of diethylaminoethyl (DEAE) cottons by reacting with cotton cellulose in the presence of sodium hydroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylamine hydrochloride acts as a precursor of atrazine and lysergic acid diethylamide. It is also used as a reactant in the production of dyes and pharmaceutical compounds such as ranitidine. Further, it is used in Mannich reaction with paraformaldehyde. In addition to this, it is employed in the synthesis of diethylaminoethyl (DEAE) cottons by reacting with cotton cellulose in the presence of sodium hydroxide.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

660-68-4
109.597
HDITUCONWLWUJR-UHFFFAOYSA-N
10197650
CCNCC.Cl
C4H12ClN
MFCD00012499
diethylamine hydrochloride, diethylammonium chloride, diethyl amine hydrochloride, n-ethylethanamine hydrochloride, ethanamine, n-ethyl-, hydrochloride, unii-ze9v3g1135, ethanamine, n-ethyl-, hydrochloride 1:1, diethylaminehydrochloride, diethylamine hcl, diethyl amine hcl
N-ethylethanamine;hydrochloride

Specifications

660-68-4
320°C to 330°C
MFCD00012499
Hygroscopic
diethylamine hydrochloride, diethylammonium chloride, diethyl amine hydrochloride, n-ethylethanamine hydrochloride, ethanamine, n-ethyl-, hydrochloride, unii-ze9v3g1135, ethanamine, n-ethyl-, hydrochloride 1:1, diethylaminehydrochloride, diethylamine hcl, diethyl amine hcl
HDITUCONWLWUJR-UHFFFAOYSA-N
N-ethylethanamine;hydrochloride
10197650
99%
227°C to 230°C
C4H12ClN
3590084
14,3111
Soluble in water.
CCNCC.Cl
109.597
109.6
Diethylamine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 211-541-9

missing translation for 'rtecsNumber' : IA3084000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only