Learn More
Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals
£86.13 - £961.79
Chemical Identifiers
CAS | 38078-09-0 |
---|---|
Molecular Formula | C4H10F3NS |
Molecular Weight (g/mol) | 161.18 |
MDL Number | MFCD00000363 |
InChI Key | CSJLBAMHHLJAAS-UHFFFAOYSA-N |
Synonym | diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4 |
PubChem CID | 123472 |
IUPAC Name | N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine |
SMILES | CCN(CC)S(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10206610
|
Thermo Scientific Acros
216110050 |
5 g | Plastic bottle |
£86.13
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10286650
|
Thermo Scientific Acros
216110250 |
25 g | Plastic bottle |
£344.72
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10318533
|
Thermo Scientific Acros
216111000 |
100 g | Plastic bottle |
£961.79
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
38078-09-0 | |
161.18 | |
CSJLBAMHHLJAAS-UHFFFAOYSA-N | |
123472 | |
CCN(CC)S(F)(F)F |
C4H10F3NS | |
MFCD00000363 | |
diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4 | |
N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine |
Specifications
38078-09-0 | |
1.3160g/mL | |
23°C | |
95% | |
(C2H5)2NSF3 | |
MFCD00000363 | |
15, 3124 | |
diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4 | |
CSJLBAMHHLJAAS-UHFFFAOYSA-N | |
N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine | |
123472 | |
95% | |
Diethylaminosulfur trifluoride |
Brown to Yellow | |
30.0°C to 32.0°C (3.0 mmHg) | |
Authentic | |
C4H10F3NS | |
1.4100 to 1.4160 | |
06,183; 10,142; 12,183; 13,110; 16,128 | |
1.316 | |
Solubility in water: decomposes. Other solubilities: soluble in benzene,thf,ether,diglyme and,dichloroethylene | |
CCN(CC)S(F)(F)F | |
161.18 | |
161.18 | |
Fuming Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapour.
Harmful in contact with skin.
Harmful if inhaled.
Reacts violently with water.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open f
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 253-771-2
- Diethylaminosulfur trifluoride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Self-reactive substance or mixture Type B
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H241-Heating may cause a fire or explosion.
- H226-Flammable liquid and vapour.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.