missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl (2-oxopropyl)phosphonate, 96%, Thermo Scientific Chemicals
For C-C Bond Formation (Olefination)
£70.40 - £250.00
Chemical Identifiers
CAS | 1067-71-6 |
---|---|
Molecular Formula | C7H19O4P |
Molecular Weight (g/mol) | 198.2 |
MDL Number | MFCD00044728 |
InChI Key | RSAFKRSMGOSHRK-UHFFFAOYSA-N |
Synonym | diethyl 2-oxopropyl phosphonate, diethyl acetonylphosphonate, diethyl 2-oxopropylphosphonate, acetonylphosphonic acid diethyl ester, 2-oxopropyl phosphonic acid diethyl ester, phosphonic acid, 2-oxopropyl-, diethyl ester, aurora ka-1476, ksc490a4t, diethyl acetylmethylphosphonate |
PubChem CID | 14028 |
IUPAC Name | 1-diethoxyphosphorylpropan-2-one |
SMILES | CCOP(=O)(CC(=O)C)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10764821
|
Thermo Scientific Acros
123690050 |
5 g | Glass bottle |
£70.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10607294
|
Thermo Scientific Acros
123690250 |
25 g | Glass bottle |
£250.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)Chemical Identifiers
1067-71-6 | |
198.2 | |
RSAFKRSMGOSHRK-UHFFFAOYSA-N | |
14028 | |
CCOP(=O)(CC(=O)C)OCC |
C7H19O4P | |
MFCD00044728 | |
diethyl 2-oxopropyl phosphonate, diethyl acetonylphosphonate, diethyl 2-oxopropylphosphonate, acetonylphosphonic acid diethyl ester, 2-oxopropyl phosphonic acid diethyl ester, phosphonic acid, 2-oxopropyl-, diethyl ester, aurora ka-1476, ksc490a4t, diethyl acetylmethylphosphonate | |
1-diethoxyphosphorylpropan-2-one |
Specifications
1067-71-6 | |
1.1100g/mL | |
>110°C | |
96% | |
CH3COCH2P(=O)(OCH2CH3)2 | |
MFCD00044728 | |
1.11 | |
RSAFKRSMGOSHRK-UHFFFAOYSA-N | |
1-diethoxyphosphorylpropan-2-one | |
14028 | |
96% | |
Diethyl (2-oxopropyl)phosphonate |
Colorless to Yellow | |
98.0°C (1.0 mmHg) | |
Authentic | |
C7H19O4P | |
1.4325 to 1.4355 | |
13,255 | |
diethyl 2-oxopropyl phosphonate, diethyl acetonylphosphonate, diethyl 2-oxopropylphosphonate, acetonylphosphonic acid diethyl ester, 2-oxopropyl phosphonic acid diethyl ester, phosphonic acid, 2-oxopropyl-, diethyl ester, aurora ka-1476, ksc490a4t, diethyl acetylmethylphosphonate | |
CCOP(=O)(CC(=O)C)OCC | |
198.2 | |
198.2 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
RUO – Research Use Only