missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol dibutyl ether, 99+%, pure, Thermo Scientific Chemicals

Product Code. 11914741
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-73-2
Molecular Formula C12H26O3
Molecular Weight (g/mol) 218.34
InChI Key KZVBBTZJMSWGTK-UHFFFAOYSA-N
Synonym diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl
PubChem CID 8210
IUPAC Name 1-[2-(2-butoxyethoxy)ethoxy]butane
SMILES CCCCOCCOCCOCCCC

Specifications

Melting Point -60.0°C
Density 0.8800g/mL
Boiling Point 256.0°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula [CH3(CH2)3OCH2CH2]2O
Refractive Index 1.4225 to 1.4245
Quantity 2.5 L
Specific Gravity 0.88
Solubility Information Solubility in water: moderate soluble (3g/L at 20°C). Other solubilities: soluble in chloroform
Viscosity 2.39 mPa.s (20°C)
Formula Weight 218.34
Percent Purity 99+%
Grade Pure
Physical Form Liquid
Chemical Name or Material Diethylene glycol dibutyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'