Learn More
Chemical Identifiers
CAS | 111-40-0 |
---|---|
Molecular Formula | C4H13N3 |
Molecular Weight (g/mol) | 103.17 |
MDL Number | MFCD00008171 |
InChI Key | RPNUMPOLZDHAAY-UHFFFAOYSA-N |
Synonym | diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine |
PubChem CID | 8111 |
ChEBI | CHEBI:30629 |
IUPAC Name | N'-(2-aminoethyl)ethane-1,2-diamine |
SMILES | NCCNCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384489
|
Thermo Scientific Alfa Aesar
A11033.AE |
100 mL |
£24.42
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11408873
|
Thermo Scientific Alfa Aesar
A11033.AP |
500 mL |
£31.46
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11418873
|
Thermo Scientific Alfa Aesar
A11033.0F |
2500 mL |
£101.51
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.
Solubility
Miscible with water and ethanol. Immiscible with ether.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and copper.
Chemical Identifiers
111-40-0 | |
103.17 | |
RPNUMPOLZDHAAY-UHFFFAOYSA-N | |
8111 | |
N'-(2-aminoethyl)ethane-1,2-diamine |
C4H13N3 | |
MFCD00008171 | |
diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine | |
CHEBI:30629 | |
NCCNCCN |
Specifications
111-40-0 | |
0.952 | |
94°C (201°F) | |
C4H13N3 | |
1.483 | |
UN2079 | |
Air Sensitive | |
Miscible with water and ethanol. Immiscible with ether. | |
NCCNCCN | |
103.17 | |
CHEBI:30629 | |
99% |
-40°C | |
200°C to 204°C | |
Ammonia-like | |
H2NCH2CH2NHCH2CH2NH2 | |
MFCD00008171 | |
605314 | |
diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine | |
RPNUMPOLZDHAAY-UHFFFAOYSA-N | |
N'-(2-aminoethyl)ethane-1,2-diamine | |
8111 | |
103.17 | |
Diethylenetriamine |
Safety and Handling
GHS H Statement
H314-H302-H312-H317
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
P210-P235-P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H227-H302+H312-H314-H317-H330-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DIETHYLENETRIAMINE
missing translation for 'einecsNumber' : 203-865-4
missing translation for 'rtecsNumber' : IE1225000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Diethylenetriamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Diethylenetriamine
RUO – Research Use Only