missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriaminepentaacetic acid, 98+%

Product Code. 10550551 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 10550551

Brand: Thermo Scientific Acros 114320010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67-43-6 | C14H23N3O10 | 393.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-43-6
Molecular Formula C14H23N3O10
Molecular Weight (g/mol) 393.34
InChI Key QPCDCPDFJACHGM-UHFFFAOYSA-N
Synonym pentetic acid, diethylenetriaminepentaacetic acid, dtpa, detapac, complexon v, detarex, perma kleer, titriplex v, monaquest cai, hamp-ex acid
PubChem CID 3053
ChEBI CHEBI:35739
IUPAC Name 2-[bis[2-[bis(carboxymethyl)amino]ethyl]amino]acetic acid
SMILES C(CN(CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O

Specifications

Melting Point 220.0°C
Color White
Density 1.5600g/mL
Flash Point 200°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Quantity 1 kg
Beilstein 04,III,119
Merck Index 15,7237
Specific Gravity 1.56
Solubility Information Solubility in water: 5g/L (20°C), 13g/L (50°C), 48g/L (80°C). Other solubilities: soluble in alkali
Formula Weight 393.34
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Diethylenetriaminepentaacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethylenetriaminepentaacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.