missing translation for 'onlineSavingsMsg'
Learn More
Diethylenetriaminepentaacetic acid dianhydride, 95%, Thermo Scientific Chemicals Product Code.: 11328186

Diethylenetriaminepentaacetic acid dianhydride, 95%, Thermo Scientific Chemicals

Product Code. 11328186
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11328186

Brand: Thermo Scientific Alfa Aesar L15178.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Diethylenetriaminepentaacetic acid dianhydride is a reagent used for introducing a strong chelator. It is employed in the synthesis of macrocyclic ligands and lanthanide complexes that are useful as MRI enhancers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylenetriaminepentaacetic acid dianhydride is a reagent used for introducing a strong chelator. It is employed in the synthesis of macrocyclic ligands and lanthanide complexes that are useful as MRI enhancers.

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is-20°C. It is sensitive to moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 23911-26-4
Molecular Formula C14H19N3O8
Molecular Weight (g/mol) 357.319
MDL Number MFCD00010697
InChI Key RAZLJUXJEOEYAM-UHFFFAOYSA-N
Synonym dtpa-anhydride, cyclic dtpa anhydride, 2-bis 2-2,6-dioxomorpholino ethyl amino acetic acid, unii-wnm9rlh5x9, wnm9rlh5x9, diethylenetriaminepentaacetic dianhydride, diethylenetriaminepentaacetic acid dianhydride, cdtpaa, glycine, n,n-bis 2-2,6-dioxo-4-morpholinyl ethyl, bis 2-2,6-dioxomorpholin-4-yl ethyl amino acetic acid
PubChem CID 100825
IUPAC Name 2-[bis[2-(2,6-dioxomorpholin-4-yl)ethyl]amino]acetic acid
SMILES C1C(=O)OC(=O)CN1CCN(CCN2CC(=O)OC(=O)C2)CC(=O)O

Specifica

Melting Point ∽190°C (decomposition)
Quantity 5 g
Beilstein 1230954
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 357.32
Percent Purity 95%
Chemical Name or Material Diethylenetriaminepentaacetic acid dianhydride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10237926 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL15178.06,L15178.03
Lot Number10241449 Certificate TypeCertificate of Analysis Date18/07/2022 Catalog NumberL15178.06,L15178.03
Lot Number10201454 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL15178.06,L15178.03
Lot Number10156165 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL15178.06,L15178.03
Lot Number10182067 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL15178.06,L15178.03

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
Diethylenetriaminepentaacetic acid dianhydride, 95%, Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato