missing translation for 'onlineSavingsMsg'
Learn More

Diethylphenylphosphine, 97%, Thermo Scientific Chemicals

Product Code. 11391228
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Diethylphenylphosphine is used as a catalyst for selective cross-dimerization, carboxyl migration reactions, selective hydrogenation, asymmetric induction by chiral diphosphines in ring contraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1605-53-4
Molecular Formula C10H15P
Molecular Weight (g/mol) 166.20
MDL Number MFCD00015172
InChI Key LVTCZSBUROAWTE-UHFFFAOYSA-N
Synonym diethylphenylphosphine, diethylphenylphosphene, diethyl phenyl phosphane, phosphine, diethylphenyl, diethyl phenyl phosphine, phenyldiethylphosphine, diethylphenyl phosphine, acmc-209dl7
PubChem CID 74149
IUPAC Name diethyl(phenyl)phosphane
SMILES CCP(CC)C1=CC=CC=C1

Specifications

Density 0.954 g/mL
Boiling Point 220°C
Flash Point 66°C (150°F)
Odor Pungent
Refractive Index 1.5475
Quantity 25 g
UN Number UN3278
Sensitivity Air and moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 166.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethylphenylphosphine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'