Learn More
Diethylphenylphosphine, 97%, Thermo Scientific Chemicals
£31.54 - £392.08
Chemical Identifiers
CAS | 1605-53-4 |
---|---|
Molecular Formula | C10H15P |
Molecular Weight (g/mol) | 166.20 |
MDL Number | MFCD00015172 |
InChI Key | LVTCZSBUROAWTE-UHFFFAOYSA-N |
Synonym | diethylphenylphosphine, diethylphenylphosphene, diethyl phenyl phosphane, phosphine, diethylphenyl, diethyl phenyl phosphine, phenyldiethylphosphine, diethylphenyl phosphine, acmc-209dl7 |
PubChem CID | 74149 |
IUPAC Name | diethyl(phenyl)phosphane |
SMILES | CCP(CC)C1=CC=CC=C1 |
Description
Diethylphenylphosphine is used as a catalyst for selective cross-dimerization, carboxyl migration reactions, selective hydrogenation, asymmetric induction by chiral diphosphines in ring contraction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1605-53-4 | |
166.20 | |
LVTCZSBUROAWTE-UHFFFAOYSA-N | |
74149 | |
CCP(CC)C1=CC=CC=C1 |
C10H15P | |
MFCD00015172 | |
diethylphenylphosphine, diethylphenylphosphene, diethyl phenyl phosphane, phosphine, diethylphenyl, diethyl phenyl phosphine, phenyldiethylphosphine, diethylphenyl phosphine, acmc-209dl7 | |
diethyl(phenyl)phosphane |
Specifications
1605-53-4 | |
220°C | |
Pungent | |
1.5475 | |
UN3278 | |
diethylphenylphosphine, diethylphenylphosphene, diethyl phenyl phosphane, phosphine, diethylphenyl, diethyl phenyl phosphine, phenyldiethylphosphine, diethylphenyl phosphine, acmc-209dl7 | |
CCP(CC)C1=CC=CC=C1 | |
166.20 | |
166.21 | |
Liquid |
0.954g/mL | |
66° (150°F) | |
C10H15P | |
MFCD00015172 | |
Air & Moisture Sensitive | |
LVTCZSBUROAWTE-UHFFFAOYSA-N | |
diethyl(phenyl)phosphane | |
74149 | |
97% | |
Diethylphenylphosphine |
Safety and Handling
GHS H Statement
H331-H227-H302-H312
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
- Diethylphenylphosphine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P363-Wash contaminated clothing before reuse.
- P321-Specific treatment (see on this label).
- P330-Rinse mouth.
- P501-Dispose of contents/container to .
RUO – Research Use Only