missing translation for 'onlineSavingsMsg'
Learn More

Diethyltin dichloride, Thermo Scientific Chemicals

Product Code. 11304849
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as organic intermediates and active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 866-55-7
Molecular Formula C4H10Cl2Sn
Molecular Weight (g/mol) 247.734
MDL Number MFCD00013609
InChI Key YFAXVVMIXZAKSR-UHFFFAOYSA-L
Synonym diethyltin dichloride, diethyldichlorotin, diethyltindichloride, dichlorodiethyltin, diethyldichlorostannane, tin, dichlorodiethyl, stannane, dichlorodiethyl, diethyltin chloride, tin, diethyl-, dichloride, unii-pga900d045
PubChem CID 91491
IUPAC Name dichloro(diethyl)stannane
SMILES CC[Sn](CC)(Cl)Cl

Specifications

Melting Point 84°C
Color White
Boiling Point 220°C
Linear Formula (C2H5)2SnCl2
Quantity 2 g
Sensitivity Moisture sensitive
Solubility Information Partly soluble in water((0.023g/L) (25°C),Calc).
Formula Weight 247.72
Physical Form Crystalline
Chemical Name or Material Diethyltin dichloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'