missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylzinc, 1.5M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 377318000
534.30 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethylzinc | |
1.5M Solution in Toluene | |
C4H10Zn | |
MFCD00009021 | |
0.9130g/mL | |
diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn | |
HQWPLXHWEZZGKY-UHFFFAOYSA-N | |
diethylzinc | |
11185 | |
AcroSeal™ Glass Bottle | |
800 mL |
Colorless | |
557-20-0, 108-88-3 | |
(C2H5)2Zn | |
1.4 to 1.6M | |
15, 3151 | |
Solubility in water: reacts violently. Other solubilities: miscible with most hydrocarbons | |
CC[Zn]CC | |
123.50 | |
123.5 | |
0.913 | |
Liquid |
RUO – Research Use Only