missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylzinc, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
Packaging:
Cylinder
Description
It is used as an important reagent in organic chemistry. It is used in organic synthesis as a source of the ethyl carbanion in addition reactions to carbonyl groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS | 557-20-0 |
Melting Point | -28°C |
Color | Colorless |
Density | 1.18 g/mL |
Boiling Point | 124°C |
Flash Point | -18°C (-1°F) |
Packaging | Cylinder |
Molecular Formula | C4H10Zn |
Linear Formula | Zn(C2H5)2 |
MDL Number | MFCD00009021 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'