missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: m Enterococcus Agar
For isolating and enumerating enterococci in water and other materials by membrane filtration or pour plate technique
£194.00
Specifications
Certifications/Compliance | CE, ISO, SMWW |
---|---|
Content And Storage | <30°C |
Form | Dehydrated |
Media Type | m Enterococcus Agar |
Product Type | Agar |
Description
- Formulation: Tryptose 20g/L, Yeast extract 5g/L, Dextrose 2g/L, Dipotassium Phosphate 4g/L, Sodium Azide 0.4g/L, Agar 10g/L, 2,3,5-Triphenyl Tetrazolium Chloride 0.1g/L
Certifications
CE, ISO, SMWW
Specifications
CE, ISO, SMWW | |
Dehydrated | |
Agar | |
For isolating and enumerating enterococci in Water and other materials by membrane filtration or pour plate technique |
<30°C | |
m Enterococcus Agar | |
Tryptose 20g/L, Yeast extract 5g/L, Dextrose 2g/L, Dipotassium Phosphate 4g/L, Sodium Azide 0.4g/L, Agar 10g/L, 2,3,5-Triphenyl Tetrazolium Chloride 0.1g/L | |
Bottle |
Safety and Handling
missing translation for 'shelfLife' : 1825 Days
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.
Supplemental information
- MIXTURE LIST-Contains : sodium azide