missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Membrane Fecal Coliform (mFC) Agar
For detection and enumeration of fecal coliforms by the membrane filter technique at elevated temperatures
£258.00
Spezifikation
Certifications/Compliance | AOAC, CCAM, EPA, SMWW |
---|---|
Content And Storage | Refrigerate |
Form | Dehydrated |
Media Type | mFC Agar |
Product Type | Agar |
Beschreibung
- Use with Rosolic Acid
- Formulation: Tryptose 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 3g/L, Lactose 12.5g/L, Bile Salts No. 3 1.5g/L, Sodium Chloride 5g/L, Agar 15g/L, Aniline blue 0.1g/L
Certifications
AOAC, CCAM, EPA, SMWW
Spezifikation
AOAC, CCAM, EPA, SMWW | |
Dehydrated | |
Agar | |
For Rosolic Acid in cultivating and enumerating fecal Coliforms by the membrane filter technique at elevated temperatures |
Refrigerate | |
mFC Agar | |
Tryptose 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 3g/L, Lactose 12.5g/L, Bile Salts No. 3 1.5g/L, Sodium Chloride 5g/L, Agar 15g/L, Aniline blue 0.1g/L | |
Bottle |
Sicherheit und Handhabung
missing translation for 'shelfLife' : 1460 Days
Signalwort
Gefahrenkategorie
Gefahrenhinweis
Sicherheitshinweis
Ergänzende Informationen
- Warning
Gefahrenkategorie
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Gefahrenhinweis
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Sicherheitshinweis
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Ergänzende Informationen
- MIXTURE LIST-Contains : bile salts No. 3