missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Noble Agar
Used as a solidifying agent essentially free of impurities. Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.
£637.00
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Concentration | 1.5% |
Content And Storage | <30°C |
Additive | Noble Agar |
Form | Dehydrated |
Description
Used as a solidifying agent essentially free of impurities. Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.
- Solidifying agent
- Free of impurities
Applications:
- Used for electrophoretic and nutritional procedures
Specifications
ISO/DIS, 11014 | |
<30°C | |
Dehydrated | |
Bottle |
1.5% | |
Noble Agar | |
Preparing microbiological culture media, for Electrophoretic and nutritional procedures |
Safety and Handling
missing translation for 'shelfLife' : 1825 Days