missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar
For cultivation of Yeasts and molds. BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar is supplied in dehydrated format.
£432.00 - £467.00
Specifications
Certifications/Compliance | ISO/DIS, 11014, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, USP |
---|---|
Content And Storage | 2°C to 30°C |
Form | Dehydrated |
Media Type | Potato Dextrose Agar |
Product Type | Agar |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
11764985
|
BD
213200 |
Pail | 2000 g |
£467.00
2kg |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
10105824
|
BD
213400 |
Bottle | 500 g |
£432.00
500g |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
Description
- Dehydrated format
- Culture media
- DNR (Latex) free
Specifications
ISO/DIS, 11014, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, USP | |
Dehydrated | |
Agar |
2°C to 30°C | |
Potato Dextrose Agar | |
Isolation of Yeasts and molds |