Learn More
BD Difco™ Disposable Inoculating Needles and Loops
Sterile, disposable inoculating needles do not require flaming
£53.90 - £95.40
Product Code | Brand | Color | Volume | Quantity | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Volume | Quantity | Product Type | Price | Quantity & Availability | |||
10492683
|
BD
220214 |
Light Green | 1 μL | 250/Pk. | Inoculating Loop |
£95.40
Pack of 250 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||
10389822
|
BD
220216 |
Light Blue | 10 μL | 250/Pk. | Inoculating Loop |
£53.90
Pack of 250 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
|||
Description
- Made from a unique plastic resin for controlled flexibility
- Resist cutting the agar surface, yet provide sufficient firmness to pick stubborn colonies
- Free of lubricants, oils and electrostatic charges
- Color coded
Specifications
Plastic Resin | |
Sterile | |
Single-use |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BD Difco™ Disposable Inoculating Needles and Loops