missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Nutrient Agar

Product Code. 11728223
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11728223

Brand: BD 213000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For the cultivation of bacteria and for the enumeration of organisms in water, sewage, feces and other materials.

  • Beef extract provides carbohydrates, vitamins, nitrogen and salts
  • Peptones provide a source of nitrogen
  • Agar is the solidifying agent
  • Consists of peptone, beef extract and agar
  • Relatively simple formulation provides the nutrients necessary for the replication of a large number of microorganisms that are not excessively fastidious
TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.