missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Nutrient Broth

Product Code. 10679125
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10679125

Brand: BD 234000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

General purpose medium for the cultivation of microorganisms with nonexacting nutritional requirements

TRUSTED_SUSTAINABILITY

Specifications

Description Nutrient Broth
Form Dehydrated
For Use With (Application) General purpose medium for the cultivation of microorganisms with non-exacting nutritional requirements
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.