missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Poly Antisera
Click to view available options
Product Type:
Shigella
Unit Size:
3mL
5mL
Each
Description
Salmonella poly antiserum
- Febrile control antisera
- Helps in the detection of diseases such as salmonellosis, brucellosis and rickettsial diseases
- Qualitative and semi-quantitative slide or tube agglutination tests for homologous antibodies
- Used along with febrile antigens as positive or negative controls
- For identification of morphologically and biochemically identified Shigella species
- Can be used only in slide agglutination test
- Lyophilized, polyclonal rabbit antisera
Specifications
Specifications
| Certifications/Compliance | For in vitro diagnostic use |
| Product Type | Shigella |
| Quantity | 3 mL |
Safety and Handling
Product Identifier
- Vial Shigella Antis Poly A Lyophilized
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sodium Azide
For in vitro diagnostic use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction