missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Sabouraud Dextrose Agar
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging Type:
Bottle
Pail
Unit Size:
10kg
500g
Each
Description
Difco™ Sabouraud Dextrose Agar is a dehydrated culture media used in qualitative procedures for cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes.
- The addition of antimicrobics is a modification designed to increase bacterial inhibition.
- The low pH of approximately 5.6 is favorable for the growth of fungi and slightly inhibitory to contaminating bacteria in clinical specimens.
- General Chapters 61 and 62 of the USP describe test methods for using Sabouraud Dextrose Agar when performing the microbial enumeration tests and tests for isolating Candida albicans from nonsterile pharmaceutical products.
Specifications
Specifications
| Certifications/Compliance | BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP |
| Color | Beige |
| Content And Storage | 2 to 25°C |
| Form | Dehydrated |
| Media Type | Sabouraud Dextrose Agar |
| Product Type | Agar |
| Formulation | Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 5g/L, Dextrose 40g/L, Agar 15g/L |
| For Use With (Application) | Cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes |
| Packaging Type | Pail |
| Quantity | 10 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction