Learn More
BD Difco™ Select APS LB Broth Base
Brand:  BD 292438
Additional Details : Weight : 1.00000kg
Product Code. 15992322
  • £137.00 / Each
Estimated Shipment: 06-08-2024
to see stock.
Add to basket
Specifications
Select APS LB Broth Base
Powder
Microbiology Media
500 g
Beige
Dehydrated
Propagation and maintenance of E. coli for molecular biology.