Learn More
Diffraction Grating Demonstration Slide
Brand: N/A
Code : 03
Additional Details : Weight : 0.00720kg
Specifications
Diffraction grating demonstration slide | |
3 rulings of 100,300,600 lines/mm | |
1 Ea. |